Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-Ethylbutyryl chloride 99%

Supplier: Origen Biomedical, Inc
Danger

B23774-14 B23774-22
AAB23774-14EA 114 USD InStock
AAB23774-14 AAB23774-22
2-Ethylbutyryl chloride 99%
2-Ethylbutyryl chloride
Purity: 99% CAS No.: 2736-40-5 Molecular Formula: (CH3CH2)2CHCOCl Formula Weight: 134.61 Flash Point: 33°(91°F) Refractive Index: 1.4250 Odor: Acrid Boiling Point: 137-139° UN No.: 3094 MDL No.: MFCD00018812


Formula: C₆H₁₁ClO
MW: 134.61 g/mol
Boiling Pt: 137…139 °C
Density: 0.980
Flash Pt: 33 °C (91 °F)
Storage Temperature: Ambient
MDL Number: MFCD00018812
CAS Number: 2736-40-5
EINECS: 220-359-9
UN: 2920
ADR: 8 II
Order Now

 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty