Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

3-Pyridyloxyacetic acid 98%

Supplier: Origen Biomedical, Inc
Warning

B24780-03 B24780-06 B24780-14
AAAB24780-03EA 87.29 USD InStock
AAAB24780-03 AAAB24780-06 AAAB24780-14
3-Pyridyloxyacetic acid 98%
3-Pyridyloxyacetic acid
Purity: 98% CAS No.: 86649-57-2 Molecular Formula: C7H7NO3 Formula Weight: 153.14 Melting Point: 210-212° MDL No.: MFCD04039772


Formula: C₇H₇NO₃
MW: 153.14 g/mol
Melting Pt: 210…212 °C
Storage Temperature: Ambient
MDL Number: MFCD04039772
CAS Number: 86649-57-2
Order Now

 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty