Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

4-Bromobenzenediazonium tetrafluoroborate 96%

Supplier: Origen Biomedical, Inc
Warning

B25670-06 B25670-14
AAAB25670-06EA 66.65 USD InStock
AAAB25670-06 AAAB25670-14
4-Bromobenzenediazonium tetrafluoroborate 96%
4-Bromobenzenediazonium tetrafluoroborate
Purity: 96% CAS No.: 673-40-5 Molecular Formula: C6H4BBrF4N2 Formula Weight: 270.83 Storage Temperature: 2°C to 8°C Melting Point: ca 135° dec. UN No.: 1759 MDL No.: MFCD00011894


Formula: C₆H₄BBrF₄N₂
MW: 270.83 g/mol
Melting Pt: ∼ 135 °C (dec.)
Storage Temperature: Refrigerator
MDL Number: MFCD00011894
CAS Number: 673-40-5
EINECS: 211-608-2
UN: 2923
ADR: 8 III
Order Now

 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty