Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

β-Cyclocitral 90+%

Supplier: Origen Biomedical, Inc
Warning

B25700-18 B25700-09
AAB25700-18EA 773 USD InStock
AAB25700-18 AAB25700-09
β-Cyclocitral 90+%
β-Cyclocitral
Purity: 90+% CAS No.: 432-25-7 Molecular Formula: C10H16O Formula Weight: 152.23 Flash Point: 110°(230°F) Refractive Index: 1.4980 Boiling Point: 62-63°/3mm MDL No.: MFCD00079078


Formula: C₁₀H₁₆O
MW: 152.23 g/mol
Boiling Pt: 62…63 °C/3 mm
Density: 0.944
Flash Pt: >110 °C(230 °F)
Storage Temperature: Ambient
MDL Number: MFCD00079078
CAS Number: 432-25-7
EINECS: 207-081-3
Order Now

 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty