Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

3-(Trifluoromethyl)piperidine 97%

Supplier: Origen Biomedical, Inc
Danger

H25851-03 H25851-MD
AAH25851-03EA 200 USD InStock
AAH25851-03 AAH25851-MD
3-(Trifluoromethyl)piperidine 97%
3-(Trifluoromethyl)piperidine
Purity: 97% CAS No.: 768-31-0 Molecular Formula: C6H10F3N Formula Weight: 153.15 Flash Point: 28°(83°F) Refractive Index: 1.3930 Odor: Amine-like Boiling Point: 128-130° UN No.: 2733 MDL No.: MFCD02183561


Formula: C₆H₁₀F₃N
Boiling Pt: 128…130 °C
Density: 1.16
Flash Pt: 28 °C(83 °F)
Storage Temperature: Ambient
MDL Number: MFCD02183561
CAS Number: 768-31-0
UN: 1992
ADR: "3",III
Order Now

 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty