Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

tert-Butyl-N-allylcarbamate 97%

Supplier: Origen Biomedical, Inc
Warning

H26014-09 H26014-18
AAH26014-09EA 69.2 USD InStock
AAH26014-09 AAH26014-18
tert-Butyl-N-allylcarbamate 97%
tert-Butyl-N-allylcarbamate
Purity: 97% CAS No.: 78888-18-3 Molecular Formula: C8H15NO2 Formula Weight: 157.21 Flash Point: 79°(175°F) Melting Point: 34-39° Boiling Point: 48-50°/1mm MDL No.: MFCD00191870


Formula: C₈H₁₅NO₂
Boiling Pt: 48…50 °C/1 mm
Melting Pt: 34…39 °C
Density: 0.938
Flash Pt: 79 °C(175 °F)
Storage Temperature: Ambient
MDL Number: MFCD00191870
CAS Number: 78888-18-3
Order Now

 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty