H26063-06
H26063-14
AAH26063-06EA
104
USD
InStock
AAH26063-06
AAH26063-14
1-Bromo-3-hexylbenzene 97+%
1-Bromo-3-hexylbenzene
Purity: 97+%
CAS No.: 38409-59-5
Molecular Formula: C12H17Br
Formula Weight: 241.17
Flash Point: 120°(248°F)
Refractive Index: 1.5220
Boiling Point: 90-98°/5mm
MDL No.: MFCD09909567