H26225-18
H26225-30
H26225-09
AAH26225-18EA
173
USD
InStock
AAH26225-18
AAH26225-30
AAH26225-09
4-(Methylthio)phenol 98%
4-(Methylthio)phenol
Purity: 98%
CAS No.: 1073-72-9
Molecular Formula: C7H8OS
Formula Weight: 140.21
Odor: Stench
Melting Point: 85-87°
Boiling Point: 153-156°/20mm
MDL No.: MFCD00002351