H26645-06
H26645-14
AAH26645-06EA
139.85
USD
InStock
AAH26645-06
AAH26645-14
(±)-Propranolol hydrochloride 99%
(±)-Propranolol hydrochloride
A beta-adrenergic blocker
Purity: 99%
CAS No.: 318-98-9
Molecular Formula: C16H21NO2.HCl
Formula Weight: 295.80
Melting Point: 163-165°
MDL No.: MFCD00012558