Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

1,4,7,10-Tetrakis(aminocarbonylmethyl)-1,4,7,10-tetraazacyclododecane

Supplier: Origen Biomedical, Inc
Warning

H26792-03 H26792-MD
AAH26792-03EA 298 USD InStock
AAH26792-03 AAH26792-MD
1,4,7,10-Tetrakis(aminocarbonylmethyl)-1,4,7,10-tetraazacyclododecane
1,4,7,10-Tetrakis(aminocarbonylmethyl)-1,4,7,10-tetraazacyclododecane
CAS No.: 157599-02-5 Molecular Formula: C16H32N8O4 Formula Weight: 400.48 MDL No.: MFCD09263318


Formula: C₁₆H₃₂N₈O₄
Storage Temperature: Ambient
MDL Number: MFCD09263318
CAS Number: 157599-02-5
Order Now

 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty