Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-Fluoro-5-(trifluoromethoxy)benzyl bromide 97%

Supplier: Origen Biomedical, Inc
Danger

H26795-03 H26795-06
AAH26795-03EA 124 USD InStock
AAH26795-03 AAH26795-06
2-Fluoro-5-(trifluoromethoxy)benzyl bromide 97%
2-Fluoro-5-(trifluoromethoxy)benzyl bromide
Purity: 97% CAS No.: 86256-24-8 Molecular Formula: C8H5BrF4O Formula Weight: 273.02 Refractive Index: 1.4660


Formula: C₈H₅BrF₄O
Storage Temperature: Ambient
MDL Number: MFCD04115890
CAS Number: 86256-24-8
UN: 3265
ADR: 8,II
Order Now

 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty