Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-Chloro-6-methoxy-3-quinoline carboxaldehyde 99%

Supplier: Origen Biomedical, Inc
Warning

H26973-14 H26973-06
AAH26973-14EA 227 USD InStock
AAH26973-14 AAH26973-06
2-Chloro-6-methoxy-3-quinoline carboxaldehyde 99%
2-Chloro-6-methoxy-3-quinoline carboxaldehyde
Purity: 99% CAS No.: 73568-29-3 Molecular Formula: C11H8ClNO2 Formula Weight: 221.64 Flash Point: -9°(16°F) Melting Point: 146-151° MDL No.: MFCD00160585


Formula: C₁₁H₈ClNO₂
MW: 221.64 g/mol
Melting Pt: 146…151 °C
Flash Pt: –9 °C (16 °F)
Storage Temperature: Ambient
MDL Number: MFCD00160585
CAS Number: 73568-29-3
Order Now

 

ORDER

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty