Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

1-Boc-3-fluoro-4-pyrrolidinone

Supplier: Origen Biomedical, Inc

H27082-MD H27082-03
AAH27082-MDEA 188 USD InStock
AAH27082-MD AAH27082-03
1-Boc-3-fluoro-4-pyrrolidinone
1-Boc-3-fluoro-4-pyrrolidinone
CAS No.: 845894-03-3 Molecular Formula: C9H14FNO3 Formula Weight: 203.22 MDL No.: MFCD08274459


Formula: C₉H₁₄FNO₃
Storage Temperature: Ambient
MDL Number: MFCD08274459
CAS Number: 845894-03-3
Order Now

 

ORDER

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty