Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

5-Amino-1-(3-chlorophenyl)-1H-pyrazole-4-carbonitrile 97%

Supplier: Origen Biomedical, Inc
Warning

H27287-06 H27287-03
AAH27287-06EA 121 USD InStock
AAH27287-06 AAH27287-03
5-Amino-1-(3-chlorophenyl)-1H-pyrazole-4-carbonitrile 97%
5-Amino-1-(3-chlorophenyl)-1H-pyrazole-4-carbonitrile
Purity: 97% CAS No.: 51516-68-8 Molecular Formula: C10H7ClN4 Formula Weight: 218.65 Melting Point: 183-187° MDL No.: MFCD00128298


Formula: C₁₀H₇ClN₄
MW: 218.65 g/mol
Melting Pt: 183…187 °C
Storage Temperature: Ambient
MDL Number: MFCD00128298
CAS Number: 51516-68-8
Order Now

 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty