Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

(2S,4S)-1-Boc-4-diphenylphosphino-2-(diphenylphosphinomethyl)pyrrolidine

Supplier: Origen Biomedical, Inc
Warning

H27321-03 H27321-MD
AAH27321-03EA 323 USD InStock
AAH27321-03 AAH27321-MD
(2S,4S)-1-Boc-4-diphenylphosphino-2-(diphenylphosphinomethyl)pyrrolidine
(2S,4S)-1-Boc-4-diphenylphosphino-2-(diphenylphosphinomethyl)pyrrolidine
CAS No.: 61478-28-2 Molecular Formula: C34H37NO2P2 Formula Weight: 553.62 Storage Temperature: 2°C to 8°C Melting Point: 100-103° MDL No.: MFCD01631275


Formula: C₃₄H₃₇NO₂P₂
Storage Temperature: Ambient
MDL Number: MFCD01631275
CAS Number: 61478-28-2
Order Now

 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty