H27445-09
H27445-18
AAH27445-09EA
70.5
USD
InStock
AAH27445-09
AAH27445-18
4-Hexen-3-one 95% (trans-isomers)
4-Hexen-3-one
Purity: 98%
CAS No.: 2497-21-4
Molecular Formula: C6H10O
Formula Weight: 98.15
Storage Temperature: -30°C to -10°C
Flash Point: 34°(93°F)
Refractive Index: 1.44
Odor: Characteristic
Boiling Point: 135-137°
UN No.: 1224
MDL No.: MFCD00010631