Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

3-(Benzyloxy)pyrrolidine 96%

Supplier: Origen Biomedical, Inc
Warning

H50769-03 H50769-MD
AAAH50769-03EA 232 USD InStock
AAAH50769-03 AAAH50769-MD
3-(Benzyloxy)pyrrolidine 96%
3-(Benzyloxy)pyrrolidine
Purity: 96% CAS No.: 420137-14-0 Molecular Formula: C11H15NO Formula Weight: 177.25 MDL No.: MFCD08060975


Formula: C₁₁H₁₅NO
MW: 177.247 g/mol
Storage Temperature: Ambient
MDL Number: MFCD08060975
CAS Number: 420137-14-0
Order Now

 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty