Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

4-Phenyltetrahydropyran-4-carboxaldehyde 95%

Supplier: Origen Biomedical, Inc
Warning

H50816-MD H50816-03
AAAH50816-MDEA 77.6 USD InStock
AAAH50816-MD AAAH50816-03
4-Phenyltetrahydropyran-4-carboxaldehyde 95%
4-Phenyltetrahydropyran-4-carboxaldehyde
Purity: 95% CAS No.: 66109-88-4 Molecular Formula: C12H14O2 Formula Weight: 190.24 MDL No.: MFCD08061006


Formula: C₁₂H₁₄O₂
Storage Temperature: Ambient
MDL Number: MFCD08061006
CAS Number: 66109-88-4
Order Now

 

ORDER

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty