Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

(2-Benzimidazolyl)acetonitrile 99%

Supplier: Origen Biomedical, Inc

L00827-09 L00827-18
AAAL00827-04EA 28.03 USD InStock
AAAL00827-04 AAAL00827-09 AAAL00827-18
(2-Benzimidazolyl)acetonitrile 99%
(2-Benzimidazolyl)acetonitrile
Purity: 99% CAS No.: 4414-88-4 Molecular Formula: C9H7N3 Formula Weight: 157.18 Odor: Odorless Melting Point: 206-211° Beilstein Reference: 128461 MDL No.: MFCD00005601


Formula: C₉H₇N₃
MW: 157.18 g/mol
Melting Pt: 206…211 °C
Storage Temperature: Ambient
MDL Number: MFCD00005601
CAS Number: 4414-88-4
EINECS: 224-574-9
Order Now

 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty