Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

4-Chlorobenzylideneacetone 98%

Supplier: Origen Biomedical, Inc
Warning

L05321-06 L05321-14
AAAL05321-06EA 48.5 USD InStock
AAAL05321-06 AAAL05321-14
4-Chlorobenzylideneacetone 98%
4-Chlorobenzylideneacetone
Purity: 98% CAS No.: 3160-40-5 Molecular Formula: C10H9ClO Formula Weight: 180.64 Melting Point: 55-60° Boiling Point: 145-147°/5mm MDL No.: MFCD00018790


Formula: C₁₀H₉ClO
Boiling Pt: 145…147 °C/5 mm
Melting Pt: 55…60 °C
Storage Temperature: Ambient
MDL Number: MFCD00018790
CAS Number: 3160-40-5
EINECS: 221-610-5
Order Now

 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty