L05776-03
L05776-06
AAAL05776-03EA
46.1
USD
InStock
AAAL05776-03
AAAL05776-06
2-Benzylcyclohexanone 97%
2-Benzylcyclohexanone
Purity: 97%
CAS No.: 946-33-8
Molecular Formula: C13H16O
Formula Weight: 188.27
Flash Point: 110°(230°F)
Refractive Index: 1.5360
Melting Point: 28-30°
Boiling Point: 103-105°/0.2mm
Beilstein Reference: 2046670
MDL No.: MFCD00051455