Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Ethyl-3-phenoxypropionate 98%

Supplier: Origen Biomedical, Inc

L07606-18 L07606-09
AAAL07606-18EA 220 USD InStock
AAAL07606-18 AAAL07606-09
Ethyl-3-phenoxypropionate 98%
Ethyl-3-phenoxypropionate
Purity: 98% CAS No.: 22409-91-2 Molecular Formula: C6H5OCH2CH2CO2CH2CH3 Formula Weight: 194.23 Flash Point: 110°(230°F) Refractive Index: 1.5005 Melting Point: 22-24° Boiling Point: 141-143°/11mm Beilstein Reference: 2331072 MDL No.: MFCD00026919


Formula: C₆H₅OCH₂CH₂CO₂CH₂CH₃
MW: 194.23 g/mol
Boiling Pt: 141…143 °C (11 mmHg)
Melting Pt: 22…24 °C
Density: 1.084
Flash Pt: >110 °C (230 °F)
Storage Temperature: Ambient
MDL Number: MFCD00026919
CAS Number: 22409-91-2
Order Now

 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty