Stabilization:
Stabilised with 4-tert-butylpyrocatechol (TBC) 0.1%
L07860-06
L07860-14
AAAL07860-06EA
90.7
USD
InStock
AAAL07860-06
AAAL07860-14
2,6-Dichlorostyrene 96% stabilized
2,6-Dichlorostyrene
Stab. with 0.1% 4-tert-butylcatechol
Purity: 96%
CAS No.: 28469-92-3
Molecular Formula: C8H6Cl2
Formula Weight: 173.04
Storage Temperature: 2°C to 8°C
Flash Point: 71°(159°F)
Refractive Index: 1.5740
Boiling Point: 88-90°/8mm
Beilstein Reference: 1931527
MDL No.: MFCD00000579