L08437-06
L08437-14
AAAL08437-06EA
78.7
USD
InStock
AAAL08437-06
AAAL08437-14
N-Phenyliminodiacetic acid 97%
N-Phenyliminodiacetic acid
Purity: 97%
CAS No.: 1137-73-1
Molecular Formula: C10H11NO4
Formula Weight: 209.20
Melting Point: ca 140° dec.
Beilstein Reference: 2808435