Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

N,N-Dimethyldecanamide 98%

Supplier: Origen Biomedical, Inc

L08731-14 L08731-06
AAAL08731-14EA 311 USD InStock
AAAL08731-14 AAAL08731-06
N,N-Dimethyldecanamide 98%
N,N-Dimethyldecanamide
Purity: 98% CAS No.: 14433-76-2 Molecular Formula: CH3(CH2)8CON(CH3)2 Formula Weight: 199.34 Refractive Index: 1.4540 Boiling Point: 110-111°/0.5mm Beilstein Reference: 1906042 MDL No.: MFCD00043725


Formula: C₁₂H₂₅NO
MDL Number: MFCD00043725
CAS Number: 14433-76-2
Order Now

 

ORDER

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty