L10129-03
L10129-06
AAAL10129-03EA
51.9
USD
InStock
AAAL10129-03
AAAL10129-06
Cyclohexanemethyl isothiocyanate 97%
Cyclohexanemethyl isothiocyanate
Purity: 97%
CAS No.: 52395-66-1
Molecular Formula: C8H13NS
Formula Weight: 155.26
Flash Point: 110°(230°F)
Refractive Index: 1.5360
Boiling Point: 222-223°
Beilstein Reference: 2079123
UN No.: 2922
MDL No.: MFCD00040881