L11845-06
L11845-03
AAAL11845-06EA
200
USD
InStock
AAAL11845-06
AAAL11845-03
4-Aminostyrene 97% stabilized
4-Aminostyrene
Purity: 97%
CAS No.: 1520-21-4
Molecular Formula: C8H9N
Formula Weight: 119.17
Storage Temperature: 2°C to 8°C
Flash Point: 110°(230°F)
Refractive Index: 1.6260
Odor: Odorless
Melting Point: 23-24°
Boiling Point: 92-94°/4mm
Beilstein Reference: 2204173
MDL No.: MFCD00015329