Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

(-)-Cotinine 98%

Supplier: Origen Biomedical, Inc
Warning

L11873-03 L11873-MD
AAAL11873-03EA 199 USD InStock
AAAL11873-03 AAAL11873-MD
(-)-Cotinine 98%
(-)-Cotinine
Purity: 98% CAS No.: 486-56-6 Molecular Formula: C10H12N2O Formula Weight: 176.22 Storage Temperature: 2°C to 8°C Flash Point: 110°(230°F) Melting Point: 34-39° Boiling Point: 145-147°/3mm Beilstein Reference: 83099 Merck Reference: 14,2553 Optical Rotation: -20° (c=1 in ethanol) MDL No.: MFCD00077696


Formula: C₁₀H₁₂N₂O
Boiling Pt: 145…147 °C/3 mm
Melting Pt: 34…39 °C
Flash Pt: >110 °C(230 °F)
Storage Temperature: Ambient
MDL Number: MFCD00077696
CAS Number: 486-56-6
EINECS: 207-634-9
Merck Index: 14,2553
Order Now
Specification Test Results
Optical rotation -20° (c=1 in ethanol)

 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty