L12270-06
L12270-14
AAAL12270-06EA
53.7
USD
InStock
AAAL12270-06
AAAL12270-14
Malonic acid dihydrazide 99%
Malonic acid dihydrazide
Purity: 99%
CAS No.: 3815-86-9
Molecular Formula: C3H8N4O2
Formula Weight: 132.12
Melting Point: 152-156°
Beilstein Reference: 1072337
MDL No.: MFCD00041268