L13601-06
L13601-14
AAAL13601-06EA
70.6
USD
InStock
AAAL13601-06
AAAL13601-14
O-Benzylhydroxylamine 96%
O-Benzylhydroxylamine
Fieser: 14,28 16,22
Purity: 96%
CAS No.: 622-33-3
Molecular Formula: C7H9NO
Formula Weight: 123.15
Refractive Index: 1.5410
Boiling Point: 118-119°/30mm
Beilstein Reference: 1906691
UN No.: 2735
MDL No.: MFCD00221709