L14659-06
L14659-03
AAAL14659-06EA
188
USD
InStock
AAAL14659-06
AAAL14659-03
Cycloheptanemethylamine ≥98%
Cycloheptanemethylamine
Purity: 98+%
CAS No.: 4448-77-5
Molecular Formula: C8H17N
Formula Weight: 127.23
Flash Point: 70°(158°F)
Refractive Index: 1.4740
Boiling Point: 98-101°/35mm
Beilstein Reference: 2958734
UN No.: 2735
MDL No.: MFCD00021674