L15107-AE
L15107-AC
AAAL15107-AEEA
114
USD
InStock
AAAL15107-AE
AAAL15107-AC
Diethylmethoxyborane 1 M in THF
Diethylmethoxyborane
CAS No.: 7397-46-8
Molecular Formula: C5H13BO
Formula Weight: 99.97
Flash Point: -6°(21°F)
Refractive Index: 1.4040
Beilstein Reference: 2231225
UN No.: 2924
MDL No.: MFCD00013240