Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Ethoxytriethylsilane 97%

Supplier: Origen Biomedical, Inc
Warning

L16656-06 L16656-14
AAAL16656-06EA 55.2 USD InStock
AAAL16656-06 AAAL16656-14
Ethoxytriethylsilane 97%
Ethoxytriethylsilane
Purity: 97% CAS No.: 597-67-1 Molecular Formula: C8H20OSi Formula Weight: 160.33 Refractive Index: 1.4140 Boiling Point: 154-155° Beilstein Reference: 1735027 UN No.: 1993 MDL No.: MFCD00054718


Formula: C₈H₂₀OSi
MW: 160.33 g/mol
Boiling Pt: 154…155 °C
Density: 0.816
Storage Temperature: Ambient
MDL Number: MFCD00054718
CAS Number: 597-67-1
UN: 1993
ADR: 3 III
Order Now

 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty