Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-Chloro-N-methoxy-N-methylacetamide 98%

Warning

L17048-14 L17048-06
AAAL17048-14EA 208.99 USD InStock
AAAL17048-14 AAAL17048-06
2-Chloro-N-methoxy-N-methylacetamide 98%
2-Chloro-N-methoxy-N-methylacetamide
Purity: 98% CAS No.: 67442-07-3 Molecular Formula: C4H8ClNO2 Formula Weight: 137.57 Storage Temperature: 2°C to 8°C Flash Point: 104°(219°F) Melting Point: 39-41° Boiling Point: 94-95°/9mm Beilstein Reference: 1924015 MDL No.: MFCD00134232


Formula: C₄H₈ClNO₂
MW: 137.56 g/mol
Boiling Pt: 94…95 °C/9 mm
Melting Pt: 39…41 °C
Flash Pt: 104 °C(219 °F)
Storage Temperature: Ambient
MDL Number: MFCD00134232
CAS Number: 67442-07-3
Order Now

 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty