Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

1,3-Bis(3-aminopropyl)tetramethyldisiloxane 94%

Supplier: Origen Biomedical, Inc
Danger

L17295-14 L17295-22
AAAL17295-14EA 79.36 USD InStock
AAAL17295-14 AAAL17295-22
1,3-Bis(3-aminopropyl)tetramethyldisiloxane 94%
1,3-Bis(3-aminopropyl)tetramethyldisiloxane
Purity: 94% CAS No.: 2469-55-8 Molecular Formula: C10H28N2OSi2 Formula Weight: 248.52 Flash Point: 91°(195°F) Refractive Index: 1.4500 Boiling Point: 132-139°/11mm Beilstein Reference: 1760876 UN No.: 2735 MDL No.: MFCD00039786


Formula: C₁₀H₂₈N₂OSi₂
MW: 248.52 g/mol
Boiling Pt: 132…139 °C/11 mm
Density: 0.897
Flash Pt: 91 °C (195 °F)
Storage Temperature: Ambient
MDL Number: MFCD00039786
CAS Number: 2469-55-8
EINECS: 219-588-7
UN: 2735
ADR: 8 III
Order Now

 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty