L20182-06
L20182-14
AAAL20182-06EA
110.99
USD
InStock
AAAL20182-06
AAAL20182-14
AAL20182-03
(1H-Imidazol-4-yl)methanol hydrochloride 98+%
(1H-Imidazol-4-yl)methanol hydrochloride
Purity: 98+%
CAS No.: 32673-41-9
Molecular Formula: C4H6N2O.HCl
Formula Weight: 134.57
Storage Temperature: 2°C to 8°C
Melting Point: 104-108°
Beilstein Reference: 3562041
MDL No.: MFCD00012697