Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Ethyl-(S)-(+)-4-cyano-3-hydroxybutyrate ≥97%, ee 98%

Supplier: Origen Biomedical, Inc
Warning

L20259-06 L20259-14
AAAL20259-06EA 94.2 USD InStock
AAAL20259-06 AAAL20259-14
Ethyl-(S)-(+)-4-cyano-3-hydroxybutyrate ≥97%, ee 98%
Ethyl-(S)-(+)-4-cyano-3-hydroxybutyrate
Purity: 97% CAS No.: 312745-91-8 Molecular Formula: C7H11NO3 Formula Weight: 157.17 Flash Point: 110°(230°F) Refractive Index: 1.4480 Boiling Point: 270° Optical Rotation: +5°(neat) MDL No.: MFCD00672878


Formula: C₇H₁₁NO₃
MW: 157.17 g/mol
Boiling Pt: 270 °C
Density: 1.114
Flash Pt: >110 °C(230 °F)
Storage Temperature: Ambient
MDL Number: MFCD00672878
CAS Number: 312745-91-8
Order Now
Specification Test Results
Optical rotation +5°(neat)

 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty