L20266-06
L20266-03
AAAL20266-06EA
445
USD
InStock
AAAL20266-06
AAAL20266-03
2,4-Difluorophenylglyoxal hydrate 95% (dry weight)
2,4-Difluorophenylglyoxal hydrate
Purity: 95%
CAS No.: 79784-36-4
Molecular Formula: C8H4F2O2.xH2O
Formula Weight: 170.12(anhy)
Melting Point: 51-54°
Beilstein Reference: 4387465
MDL No.: MFCD04038287