Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Hydriodic acid 47% stabilised ACS

Supplier: Origen Biomedical, Inc
Danger

Stabilization: 1.5% stab. with hypophosphorous acid

036490-AE 036490-AP
AA36490-AEEA 81.9 USD InStock
AA36490-AE AA36490-AP
Hydriodic acid 47% stabilised ACS
Hydriodic acid
Fieser: 1,449 6,281 7,173 8,246 9,238 10,199 15,166 18,180 21,216

CAS No.: 10034-85-2 Molecular Formula: HI Formula Weight: 127.91 Physical Form: Liquid Density: 1.50 Sensitivity: Light Sensitive Appearance: Colorless to yellow Merck Reference: 14,4776 UN No.: 1787 MDL No.: MFCD00011347


Formula: HI
MW: 127.91 g/mol
Density: 1.50
Storage Temperature: Ambient
MDL Number: MFCD00011347
CAS Number: 10034-85-2
EINECS: 233-109-9
UN: 1787
ADR: 8 II
Merck Index: 14,4776
Order Now
Specification Test Results
Chloride and bromide (as Cl) Pass test
SO4 Pass test.
Stabilizer (H3PO2) 1.5% max.
Heavy Metals (as Pb) 0.001% max.

 

ORDER

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty