Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Tetrabutylammonium sulfate 50% (w/w) in aqueous solution

Supplier: Origen Biomedical, Inc
Warning

041723-AP 041723-K4 041723-AE
AA41723-APEA 161 USD InStock
AA41723-AP AA41723-K4 AA41723-AE
Tetrabutylammonium sulfate 50% (w/w) in aqueous solution
Tetrabutylammonium sulphate
Applications: Phase transfer catalyst, neutral pH

Phase transfer catalyst, neutral pH

Phase transfer catalyst, neutral pH


Formula: C₃₂H₇₂N₂O₄S
MW: 581 g/mol
Boiling Pt: 84 °C
Density: 1.01
Storage Temperature: Ambient
MDL Number: MFCD00216632
CAS Number: 2472-88-0
EINECS: 219-593-4
Order Now

 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty