Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Nickel ≥99.8% (metals basis), powder -150+200 mesh

Supplier: Origen Biomedical, Inc
Danger

044739-22 044739-36 044739-A3
AA44739-22EA 70.17 USD InStock
AA44739-22 AA44739-36 AA44739-A3
Nickel ≥99.8% (metals basis), powder -150+200 mesh
Nickel
Purity: 99.8% CAS No.: 7440-02-0 Appearance: Gray Odor: Odorless MDL No.: MFCD00011137


Formula: Ni
Boiling Pt: 2371 °C (1013 hPa)
Melting Pt: 1455 °C
Density: 8.9 g/cm³ (25 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011137
CAS Number: 7440-02-0
EINECS: 231-111-4
UN: 3089
ADR: 4.1,II
Order Now

 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty