Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Manganese ≥99.9% (metals basis), nanopowder APS 30-50 nm

Supplier: Origen Biomedical, Inc
Warning

045508-06 045508-14
AA45508-06EA 286.24 USD InStock
AA45508-06 AA45508-14
Manganese ≥99.9% (metals basis), nanopowder APS 30-50 nm
Manganese
Purity: 99.9% CAS No.: 7439-96-5 Molecular Formula: Mn(core)/Mn3O4(shell) Physical Form: Mn3O4 layer 5-12nm UN No.: 3089 MDL No.: MFCD00011111


Formula: Mn
Storage Temperature: Ambient
MDL Number: MFCD00011111
CAS Number: 7439-96-5
EINECS: 231-105-1
UN: 3089
ADR: 4.1,III
Order Now

 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty