Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Sodium ethoxide 21% (w/w) in ethanol

Supplier: Origen Biomedical, Inc
Danger

Packaged under argon in resealable ChemSeal™ bottles.

045590-AE 045590-AP
AA45590-AEEA 53.14 USD InStock
AA45590-AE AA45590-AP
Sodium ethoxide 21% (w/w) in ethanol
Sodium ethoxide
CAS No.: 141-52-6 Molecular Formula: C2H5NaO Formula Weight: 68.05 Flash Point: 20°(68°F) Physical Form: Liquid Density: 0.868 Refractive Index: 1.3850 Sensitivity: Moisture Sensitive Beilstein Reference: 3593646 Merck Reference: 14,8613 UN No.: 3274 MDL No.: MFCD00012417


Formula: C₂H₅NaO
MW: 68.05 g/mol
Melting Pt: 260
Density: 0.868
Flash Pt: 20 °C (68 °F)
Storage Temperature: Ambient
MDL Number: MFCD00012417
CAS Number: 141-52-6
EINECS: 205-487-5
UN: 3274
ADR: 3 II
Merck Index: 14,8613
Order Now

 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty