A15175-09
A15175-18
A15175-30
AAA15175-09EA
77.76
USD
InStock
AAA15175-09
AAA15175-18
AAA15175-30
Trimethyloxonium tetrafluoroborate 96%
Trimethyloxonium tetrafluoroborate
Fieser: 1,1232 3,314 4,541 6,630 10,426 13,327
Purity: 96%
CAS No.: 420-37-1
Molecular Formula: C3H9BF4O
Formula Weight: 147.91
Storage Temperature: -30°C to -10°C
Melting Point: ca 190° dec.
Beilstein Reference: 3597303
UN No.: 3261
MDL No.: MFCD00011798