Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

1,3-Dicyclohexylurea 98%

Supplier: Origen Biomedical, Inc

B20805-14 B20805-22
AAB20805-14EA 49.6 USD InStock
AAB20805-14 AAB20805-22 AAB20805-36
1,3-Dicyclohexylurea 98%
1,3-Dicyclohexylurea
Purity: 98% CAS No.: 2387-23-7 Molecular Formula: C13H24N2O Formula Weight: 224.35 Melting Point: 230-233° MDL No.: MFCD00003829


Formula: C₁₃H₂₄N₂O
Melting Pt: 230…233 °C
Storage Temperature: Ambient
MDL Number: MFCD00003829
CAS Number: 2387-23-7
EINECS: 219-213-7
Order Now

 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty