B21245-06
B21245-14
AAB21245-06EA
120
USD
InStock
AAB21245-06
AAB21245-14
(R)-(+)-Epoxystyrene 98%
(R)-(+)-Epoxystyrene
Purity: 98%
CAS No.: 20780-53-4
Molecular Formula: C8H8O
Formula Weight: 120.15
Storage Temperature: 2°C to 8°C
Flash Point: 74°(165°F)
Refractive Index: 1.5350
Boiling Point: 194°
Beilstein Reference: 1984
Optical Rotation: +32° (neat)
UN No.: 2810
MDL No.: MFCD00066210