Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Pyruvaldehyde (methylglyoxal) 35 - 45% (w/w) in aqueous solution

Supplier: Origen Biomedical, Inc
Danger

B24664-14 B24664-36 B24664-22
AAB24664-14EA 32.88 USD InStock
AAB24664-14 AAB24664-36 AAB24664-22
Pyruvaldehyde (methylglyoxal) 35 - 45% (w/w) in aqueous solution
Pyruvaldehyde (methylglyoxal)
CAS No.: 78-98-8 Molecular Formula: C3H4O2 Formula Weight: 72.06 Refractive Index: 1.4050 Beilstein Reference: 906750 Merck Reference: 14,6081 MDL No.: MFCD00006960


Formula: C₃H₄O₂
Density: 1.2
Storage Temperature: Ambient
MDL Number: MFCD00006960
CAS Number: 78-98-8
EINECS: 201-164-8
Merck Index: 14,6081
Order Now

 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty