Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Benzylurea 98%

Supplier: Origen Biomedical, Inc

B24945-22 B24945-14
AAAB24945-22EA 91 USD InStock
AAAB24945-22 AAAB24945-14
Benzylurea 98%
Benzylurea
Purity: 98% CAS No.: 538-32-9 Molecular Formula: C6H5CH2NHCONH2 Formula Weight: 150.18 Melting Point: 146-151° Beilstein Reference: 2208083 Merck Reference: 14,1147 MDL No.: MFCD00007951


Formula: C₈H₁₀N₂O
Melting Pt: 146…151 °C
Storage Temperature: Ambient
MDL Number: MFCD00007951
CAS Number: 538-32-9
EINECS: 208-689-1
Merck Index: 14,1147
Order Now

 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty