Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2,4-Dichloro-5-methoxyaniline 98%

Supplier: Origen Biomedical, Inc
Warning

B25497-03 B25497-06 B25497-14
AAB25497-03EA 28.1 USD InStock
AAB25497-03 AAB25497-06 AAB25497-14
2,4-Dichloro-5-methoxyaniline 98%
2,4-Dichloro-5-methoxyaniline
Purity: 98% CAS No.: 98446-49-2 Molecular Formula: C7H7Cl2NO Formula Weight: 192.05 UN No.: 2810 MDL No.: MFCD00974410


Formula: C₇H₇Cl₂NO
Storage Temperature: Ambient
MDL Number: MFCD00974410
CAS Number: 98446-49-2
UN: 2811
ADR: "6.1",III
Order Now

 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty