H26066-03
H26066-09
AAH26066-03EA
22.5
USD
InStock
AAH26066-03
AAH26066-09
3-Bromo-5-fluorobenzonitrile 98%
3-Bromo-5-fluorobenzonitrile
Purity: 98%
CAS No.: 179898-34-1
Molecular Formula: C7H3BrFN
Formula Weight: 200.01
UN No.: 3439
MDL No.: MFCD04038227